3-Phenoxybenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR12558-100MG OR12558-1G OR12558-250MG
APOSOR12558-100MGEA 98.6 EUR
APOSOR12558-100MG APOSOR12558-1G APOSOR12558-250MG
3-Phenoxybenzenesulphonyl chloride
3-Phenoxybenzenesulphonyl chloride
Formel: C₁₂H₉ClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD02089483
CAS-Nummer: 252873-46-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR