3-(2-Methoxyphenoxy)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[3-(Chlorosulphonyl)phenoxy]anisole, 3-(Chlorosulphonyl)-2'-methoxydiphenyl ether

OR12567-250MG
APOSOR12567-250MGEA 174 EUR
APOSOR12567-250MG
3-(2-Methoxyphenoxy)benzenesulphonyl chloride
3-(2-Methoxyphenoxy)benzenesulphonyl chloride
Formel: C₁₃H₁₁ClO₄S
Molecular Weight: 298,742 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631872
CAS-Nummer: 521980-26-7
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR