3-(2-Methylphenoxy)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR12569-100MG
APOSOR12569-100MGEA 0 EUR
APOSOR12569-100MG
3-(2-Methylphenoxy)benzenesulphonyl chloride
3-(2-Methylphenoxy)benzenesulphonyl chloride
Formel: C₁₃H₁₁ClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD01631874
CAS-Nummer: 885950-88-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR