2-Amino-5-chlorbenzolsulfonsäure

Lieferant: Apollo Scientific
Achtung

OR12580-100MG
APOSOR12580-100MGEA 21.9 EUR
APOSOR12580-100MG
2-Amino-5-chlorbenzolsulfonsäure
2-Amino-5-chlorobenzenesulphonic acid
Formel: C₆H₆ClNO₃S
Molecular Weight: 207.64 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035776
CAS-Nummer: 133-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR