2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98+%

Lieferant: Apollo Scientific

OR12587-250MG
APOSOR12587-250MGEA 144 EUR
APOSOR12587-250MG
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98+%
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide
Formel: C₁₂H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800601
CAS-Nummer: 80047-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR