4-Nitro-o-toluonitril 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-4-nitrobenzonitril, 2-Methyl-4-nitrobenzoesäurenitril
OR12589-25G
APOSOR12589-25GEA
221
EUR
APOSOR12589-25G
4-Nitro-o-toluonitril 98%
4-Nitro-o-toluonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095378 CAS-Nummer: 89001-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...