N-Methyl-4-phenylbutylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12593-1G OR12593-5G OR12593-10G
APOSOR12593-1GEA 159 EUR
APOSOR12593-1G APOSOR12593-5G APOSOR12593-10G
N-Methyl-4-phenylbutylamine hydrochloride
N-Methyl-4-phenylbutylamine hydrochloride
Formel: C₁₁H₁₇N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08445574
CAS-Nummer: 148252-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR