N-Methyl-4-phenylbutylamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR12593-1G
OR12593-5G
OR12593-10G
APOSOR12593-1GEA
159
EUR
APOSOR12593-1G
APOSOR12593-5G
APOSOR12593-10G
N-Methyl-4-phenylbutylamine hydrochloride
N-Methyl-4-phenylbutylamine hydrochloride
Formel:
C₁₁H₁₇N·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445574 CAS-Nummer: 148252-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...