1-Isochinolincarbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinoline-1-carboxylic acid
OR12594-250MG
APOSOR12594-250MGEA
21.9
EUR
APOSOR12594-250MG
1-Isochinolincarbonsäure 98%
1-Isochinolincarbonsäure
Formel:
C₁₀H₇NO₂ Molecular Weight: 173,17 g/mol Schmelzpunkt: 164 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006901 CAS-Nummer: 486-73-7 EINECS: 207-639-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...