4-(2-Chlorphenoxy)piperidin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
OR12600-1G
APOSOR12600-1GEA
393
EUR
APOSOR12600-1G
4-(2-Chlorphenoxy)piperidin Hydrochlorid
4-(2-Chlorphenoxy)piperidin Hydrochlorid
Formel:
C₁₁H₁₅Cl₂NO Molecular Weight: 248,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684107 CAS-Nummer: 849107-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...