[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Aminomethyl)-4'-methyldiphenyl ether hydrochloride, 4-(4-Methylphenoxy)benzylamine hydrochloride

OR12604-100MG
APOSOR12604-100MGEA 21.9 EUR
APOSOR12604-100MG
[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride
[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride
Formel: C₁₄H₁₅NO·ClH
Molecular Weight: 249,736 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089411
CAS-Nummer: 262862-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR