4-Phenoxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR12606-25G
APOSOR12606-25GEA
21.9
EUR
APOSOR12606-25G
4-Phenoxybenzoesäure
4-Phenoxybenzoesäure
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214,22 g/mol Schmelzpunkt: 160…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002539 CAS-Nummer: 2215-77-2 EINECS: 218-682-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...