(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12608-100MG
APOSOR12608-100MGEA 23.2 EUR
APOSOR12608-100MG
(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride
(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride
Formel: C₁₃H₁₃Cl₂NO
Molecular Weight: 270,1544 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089410
CAS-Nummer: 262862-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR