4-[(Pyridin-2-yl)oxy]benzenesulphonyl chloride hydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[4-(Chlorosulphonyl)phenoxy]pyridine hydrochloride

OR12616-100MG
APOSOR12616-100MGEA 23.2 EUR
APOSOR12616-100MG
4-[(Pyridin-2-yl)oxy]benzenesulphonyl chloride hydrochloride 95%
4-[(Pyridin-2-yl)oxy]benzenesulphonyl chloride hydrochloride
Formel: C₁₁H₈ClNO₃S·ClH
Molecular Weight: 306.165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862556
CAS-Nummer: 1170110-04-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR