4-(Aminomethyl)-4'-methylbiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methylphenyl)benzylamine hydrochloride, (4'-Methylbiphenyl-4-yl)methylamine hydrochloride

OR12637-100MG
APOSOR12637-100MGEA 94.4 EUR
APOSOR12637-100MG
4-(Aminomethyl)-4'-methylbiphenyl hydrochloride
4-(Aminomethyl)-4'-methylbiphenyl hydrochloride
Formel: C₁₄H₁₅N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD02089428
CAS-Nummer: 1173246-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR