5-Acetylisoxazole-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-acetylisoxazole-3-carboxylate

OR1264-1G
APOSOR1264-1GEA 161 EUR
APOSOR1264-1G
5-Acetylisoxazole-3-carboxylic acid ethyl ester
5-Acetylisoxazole-3-carboxylic acid ethyl ester
Formel: C₈H₉NO₄
Molecular Weight: 183,17 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085046
CAS-Nummer: 104776-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR