4-(2-Chlorophenyl)benzylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12640-1G OR12640-250MG OR12640-50MG
APOSOR12640-1GEA 0 EUR
APOSOR12640-1G APOSOR12640-250MG APOSOR12640-50MG
4-(2-Chlorophenyl)benzylamine hydrochloride
4-(2-Chlorophenyl)benzylamine hydrochloride
Formel: C₁₃H₁₂ClN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD02089431
CAS-Nummer: 518357-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR