2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hemioxalate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Thia-6-azaspiro[3.4]octane 2,2-dioxide ethane-1,2-dioate

OR12645-100MG
APOSOR12645-100MGEA 580 EUR
APOSOR12645-100MG
2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hemioxalate
2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hemioxalate
Formel: 2C₆H₁₁NO₂S·C₂H₂O₄
Molecular Weight: 412,479 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28384160
CAS-Nummer: 1923267-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR