4-(4-Pyridyl)benzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Pyridin-4-yl)benzaldehyde

OR12650-250MG
APOSOR12650-250MGEA 21.9 EUR
APOSOR12650-250MG
4-(4-Pyridyl)benzaldehyd 98%
4-(4-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684110
CAS-Nummer: 99163-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR