3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR12654-1G
APOSOR12654-1GEA
30.6
EUR
APOSOR12654-1G
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
3',4'-Dichloro[1,1'-biphenyl]-4-carbaldehyde
Formel:
C₁₃H₈Cl₂O Molecular Weight: 251,11 g/mol Schmelzpunkt: 95…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03002047 CAS-Nummer: 50670-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...