Methyl-3-(4-(bromomethyl)phenyl)acrylate 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-(4-(bromomethyl)phenyl)acrylate, Methyl 4-(bromomethyl)cinnamate

OR12661-100G
APOSOR12661-100GEA 150 EUR
APOSOR12661-100G
Methyl-3-(4-(bromomethyl)phenyl)acrylate 96%
Methyl-3-(4-(bromomethyl)phenyl)acrylate
Formel: C₁₁H₁₁BrO₂
Molecular Weight: 255.1100 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02684186
CAS-Nummer: 946-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR