1-(3-Phenyl-5-isoxazolyl)-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Phenylisoxazol-5yl)ethanone

OR1267-500MG
APOSOR1267-500MGEA 34.8 EUR
APOSOR1267-500MG
1-(3-Phenyl-5-isoxazolyl)-1-ethanone
1-(3-Phenyl-5-isoxazolyl)-1-ethanone
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2 g/mol
Schmelzpunkt: 103…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100033
CAS-Nummer: 2048-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR