2-(Methylsulfonyl)ethanaminhydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methylsulphonyl)ethanamine hydrochloride, 2-Aminoethyl Methyl sulphone Hydrochloride, 2-Aminoethyl methyll sulphone hydrochloride, 2-(Methylsulphonyl)ethanamine Hydrochloride

OR12674-500MG OR12674-1G
APOSOR12674-500MGEA 21.9 EUR
APOSOR12674-500MG APOSOR12674-1G
2-(Methylsulfonyl)ethanaminhydrochlorid
2-(Methylsulphonyl)ethanamine hydrochloride
Formel: C₃H₁₀ClNO₂S
Molecular Weight: 159.6400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840162
CAS-Nummer: 104458-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR