3-(Phenylsulphonyl)pyrrolidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Phenyl pyrrolidin-3-yl sulphone hydrochloride
OR12677-50MG
APOSOR12677-50MGEA
23.2
EUR
APOSOR12677-50MG
3-(Phenylsulphonyl)pyrrolidine hydrochloride
3-(Phenylsulphonyl)pyrrolidine hydrochloride
Formel:
C₁₀H₁₃NO₂S·ClH Molecular Weight: 247,742 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445575 CAS-Nummer: 1003562-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...