(4-Chlorphenyl)methansulfonsäurechlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Chloro-α-toluenesulphonyl chloride
OR12681-1G
OR12681-25G
OR12681-5G
APOSOR12681-1GEA
21.9
EUR
APOSOR12681-1G
APOSOR12681-25G
APOSOR12681-5G
(4-Chlorphenyl)methansulfonsäurechlorid
(4-Chlorophenyl)methanesulphonyl chloride
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225.10 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631927 CAS-Nummer: 6966-45-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...