2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR12694-250MG OR12694-1G
APOSOR12694-250MGEA 509 EUR
APOSOR12694-250MG APOSOR12694-1G
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08445576
CAS-Nummer: 31788-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR