3,5-Dimethyl-4-isoxazolsulfonylchlorid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR1270-5G
APOSOR1270-5GEA
36.4
EUR
APOSOR1270-5G
3,5-Dimethyl-4-isoxazolsulfonylchlorid 98%
3,5-Dimethyl-4-isoxazolsulfonylchlorid
Formel:
C₅H₆ClNO₃S Molecular Weight: 195,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052554 CAS-Nummer: 80466-79-1 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...