2-(2-Chlorophenyl)-2-(morpholin-4-yl)ethylamine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[2-Amino-1-(morpholin-4-yl)ethyl]-2-chlorobenzene, 4-[2-Amino-1-(2-chlorophenyl)ethyl]morpholine

OR12703-1G
APOSOR12703-1GEA 299 EUR
APOSOR12703-1G
2-(2-Chlorophenyl)-2-(morpholin-4-yl)ethylamine 95%
2-(2-Chlorophenyl)-2-(morpholin-4-yl)ethylamine
Formel: C₁₂H₁₇ClN₂O
Molecular Weight: 240,729 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631948
CAS-Nummer: 866782-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR