2-(2-Chlorophenyl)-2-phenylethanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-Chlorobenzhydryl)methanesulphonyl chloride

OR12708-250MG OR12708-50MG
APOSOR12708-250MGEA 244 EUR
APOSOR12708-250MG APOSOR12708-50MG
2-(2-Chlorophenyl)-2-phenylethanesulphonyl chloride
2-(2-Chlorophenyl)-2-phenylethanesulphonyl chloride
Formel: C₁₄H₁₂Cl₂O₂S
Molecular Weight: 315.215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631949
CAS-Nummer: 885950-96-9
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR