[4-(tert-Butoxycarbonyl)piperazin-1-yl](4-methylphenyl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-{[4-(tert-Butoxycarbonyl)piperazin-1-yl](carboxy)methyl}toluene, 4-[Carboxy(4-methylphenyl)methyl]piperazine, N1-BOC protected
OR12724-100MG
APOSOR12724-100MGEA
21.9
EUR
APOSOR12724-100MG
[4-(tert-Butoxycarbonyl)piperazin-1-yl](4-methylphenyl)acetic acid
[4-(tert-Butoxycarbonyl)piperazin-1-yl](4-methylphenyl)acetic acid
Formel:
C₁₈H₂₆N₂O₄ Molecular Weight: 334,41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862501 CAS-Nummer: 885274-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...