2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-cyano-4,5-dimethylpyrrole

OR1274-1G
APOSOR1274-1GEA 21.9 EUR
APOSOR1274-1G
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile
Formel: C₇H₉N₃
Molecular Weight: 135,17 g/mol
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00849263
CAS-Nummer: 21392-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR