2-(4-Methoxyphenyl)-2-phenylethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12743-50MG
APOSOR12743-50MGEA 58 EUR
APOSOR12743-50MG
2-(4-Methoxyphenyl)-2-phenylethylamine hydrochloride
2-(4-Methoxyphenyl)-2-phenylethylamine hydrochloride
Formel: C₁₅H₁₇NO·ClH
Molecular Weight: 263,763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089461
CAS-Nummer: 21998-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR