2-(4-Chlorophenyl)-2-phenylethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12744-50MG
APOSOR12744-50MGEA 58 EUR
APOSOR12744-50MG
2-(4-Chlorophenyl)-2-phenylethylamine hydrochloride
2-(4-Chlorophenyl)-2-phenylethylamine hydrochloride
Formel: C₁₄H₁₄ClN·ClH
Molecular Weight: 268,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089402
CAS-Nummer: 21998-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR