4-(Imidazo[1,2-a]pyrazin-2-yl)-1,2,5-oxadiazol-3-amine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-4-(imidazo[1,2-a]pyrazin-2-yl)-1,2,5-oxadiazole

OR12765-250MG
APOSOR12765-250MGEA 171 EUR
APOSOR12765-250MG
4-(Imidazo[1,2-a]pyrazin-2-yl)-1,2,5-oxadiazol-3-amine 97%
4-(Imidazo[1,2-a]pyrazin-2-yl)-1,2,5-oxadiazol-3-amine
Formel: C₈H₆N₆O
Molecular Weight: 202,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035755
CAS-Nummer: 1031927-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR