5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-(methylsulphonyl)-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate, tert-Butyl 5,7-dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine-6-carboxylate

OR12766-250MG
APOSOR12766-250MGEA 42.2 EUR
APOSOR12766-250MG
5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected 97%
5,7-Dihydro-2-(methylsulphonyl)-6H-pyrrolo[3,4-d]pyrimidine, N6-BOC protected
Formel: C₁₂H₁₇N₃O₄S
Molecular Weight: 299,346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839781
CAS-Nummer: 365996-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR