Methyl 2-bromo-5-cyanobenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-3-(methoxycarbonyl)benzonitrile

OR12768-250MG
APOSOR12768-250MGEA 21.9 EUR
APOSOR12768-250MG
Methyl 2-bromo-5-cyanobenzoate
Methyl 2-bromo-5-cyanobenzoate
Formel: C₉H₆BrNO₂
Molecular Weight: 240,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035757
CAS-Nummer: 1031927-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR