1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: cis-4-Amino-L-proline methyl ester, N-CBZ protected hydrochloride, (2S,4S)-4-Amino-1-[(benzyloxy)carbonyl]-2-(methoxycarbonyl)pyrrolidine hydrochloride

OR12769-100MG
APOSOR12769-100MGEA 62.5 EUR
APOSOR12769-100MG
1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Formel: C₁₄H₁₈N₂O₄·ClH
Molecular Weight: 314,765 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804628
CAS-Nummer: 1212395-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR