2-Methyl-1,3-benzothiazol-6-amine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-2-methyl-1,3-benzothiazole

OR12777-1G
APOSOR12777-1GEA 65.4 EUR
APOSOR12777-1G
2-Methyl-1,3-benzothiazol-6-amine 98%
2-Methyl-1,3-benzothiazol-6-amine
Formel: C₈H₈N₂S
Molecular Weight: 164.23 g/mol
Schmelzpunkt: 121…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160076
CAS-Nummer: 2941-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR