N,O-Dimethylhydroxylamin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

OR12778-100G OR12778-1KG
APOSOR12778-100GEA 0 EUR
APOSOR12778-100G APOSOR12778-1KG
N,O-Dimethylhydroxylamin Hydrochlorid 98%
N,O-Dimethylhydroxylamin Hydrochlorid
Formel: C₂H₈ClNO
Schmelzpunkt: 112…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012485
CAS-Nummer: 6638-79-5
EINECS: 229-642-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR