3-Amino-5-bromopyridin-2(1H)-one 98%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-bromo-2-hydroxypyridine

OR12788-25G
APOSOR12788-25GEA 219 EUR
APOSOR12788-25G
3-Amino-5-bromopyridin-2(1H)-one 98%
3-Amino-5-bromopyridin-2(1H)-one
Formel: C5H5BrN2O
Molecular Weight: 189.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840438
CAS-Nummer: 98786-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR