4'-Methyl-[1,1'-biphenyl]-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR12791-1G
APOSOR12791-1GEA
1010
EUR
APOSOR12791-1G
4'-Methyl-[1,1'-biphenyl]-2-sulphonyl chloride
4'-Methyl-[1,1'-biphenyl]-2-sulphonyl chloride
Formel:
C₁₃H₁₁ClO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840035 CAS-Nummer: 173253-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...