(4-Phenoxyphenyl)essigsäure 96%

Lieferant: Apollo Scientific

OR12795-5G
APOSOR12795-5GEA 60.9 EUR
APOSOR12795-5G
(4-Phenoxyphenyl)essigsäure 96%
(4-Phenoxyphenyl)essigsäure
Formel: (C₆H₅OC₆H₅)CH₂COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00079779
CAS-Nummer: 6328-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR