4-(4-Methoxyphenoxy)piperidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR12797-100MG
APOSOR12797-100MGEA 30.6 EUR
APOSOR12797-100MG
4-(4-Methoxyphenoxy)piperidin Hydrochlorid
4-(4-Methoxyphenoxy)piperidin Hydrochlorid
Formel: C₁₂H₁₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840124
CAS-Nummer: 333954-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR