4-(2-Methoxyphenoxy)piperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12804-100MG
APOSOR12804-100MGEA 0 EUR
APOSOR12804-100MG
4-(2-Methoxyphenoxy)piperidine hydrochloride
4-(2-Methoxyphenoxy)piperidine hydrochloride
Formel: C₁₂H₁₇NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03840131
CAS-Nummer: 950649-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR