3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(tert-Butyl)-5-(chlorocarbonyl)-1-methyl-1H-pyrazole, 3-(tert-Butyl)-5-(chloroformyl)-1-methyl-1H-pyrazole
OR1281-1G
APOSOR1281-1GEA
31.9
EUR
APOSOR1281-1G
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carbonyl chloride
Formel:
C₉H₁₃ClN₂O Molecular Weight: 200,665 g/mol Siedepunkt: 117 °C (15 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084916 CAS-Nummer: 160842-62-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...