3-(2-Amino-1-hydroxyethyl)pyridine dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-2-(pyridin-3-yl)ethylamine dihydrochloride, 2-Amino-1-(pyridin-3-yl)ethan-1-ol dihydrochloride
OR12818-250MG
APOSOR12818-250MGEA
140
EUR
APOSOR12818-250MG
3-(2-Amino-1-hydroxyethyl)pyridine dihydrochloride
3-(2-Amino-1-hydroxyethyl)pyridine dihydrochloride
Formel:
C₇H₁₀N₂O·2ClH Molecular Weight: 211.089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028161 CAS-Nummer: 101012-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...