3-(4-Methylphenyl)morpholine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(p-Tolyl)morpholine hydrochloride, 4-(Morpholin-3-yl)toluene hydrochloride

OR12854-100MG
APOSOR12854-100MGEA 183 EUR
APOSOR12854-100MG
3-(4-Methylphenyl)morpholine hydrochloride
3-(4-Methylphenyl)morpholine hydrochloride
Formel: C₁₁H₁₅NO·ClH
Molecular Weight: 213,704 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840091
CAS-Nummer: 1170445-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR