3-(2-Methoxyphenyl)morpholine oxalate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Morpholin-3-yl)anisole oxalate, 3-(2-Methoxyphenyl)morpholine ethane-1,2-dioate

OR12856-25MG
APOSOR12856-25MGEA 48 EUR
APOSOR12856-25MG
3-(2-Methoxyphenyl)morpholine oxalate
3-(2-Methoxyphenyl)morpholine oxalate
Lagertemperatur: Raumtemperatur
MDL: MFCD04117745
CAS-Nummer: 1170934-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR