1,3-Oxazole-4-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxazolecarboxaldehyde, 4-Formyloxazole

OR12860-1G
APOSOR12860-1GEA 174 EUR
APOSOR12860-1G
1,3-Oxazole-4-carbaldehyde
1,3-Oxazole-4-carbaldehyde
Formel: C₄H₃NO₂
Molecular Weight: 97,07 g/mol
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179512
CAS-Nummer: 118994-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR