4-(4-Methoxybenzyl)piperidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(Piperidin-4-yl)methyl]anisole hydrochloride

OR12871-1G OR12871-250MG OR12871-5G
APOSOR12871-1GEA 198 EUR
APOSOR12871-1G APOSOR12871-250MG APOSOR12871-5G
4-(4-Methoxybenzyl)piperidine hydrochloride 95%
4-(4-Methoxybenzyl)piperidine hydrochloride
Formel: C₁₃H₁₉NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03840137
CAS-Nummer: 37581-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR